missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | MAGEB2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
MAGEB2 Polyclonal specifically detects MAGEB2 in Human samples. It is validated for Western Blot.Specifications
| MAGEB2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Cancer/testis antigen 3.2, cancer/testis antigen family 3, member 2, CT3.2MAGE-B2 antigen, DAM6MAGE XP-2 antigen, DSS/AHC critical interval MAGE superfamily 6, DSS-AHC critical interval MAGE superfamily 6, MAGE-XP-2, melanoma antigen family B, 2, melanoma-associated antigen B2, MGC26438 | |
| MAGEB2 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| O15479 | |
| 4113 | |
| Synthetic peptides corresponding to MAGEB2(melanoma antigen family B, 2) The peptide sequence was selected from the N terminal of MAGEB2. Peptide sequence AAAAGVSSTKSKKGAKSHQGEKNASSSQASTSTKSPSEDPLTRKSGSLVQ. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title