missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB18 Antibody (OTI1H2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72577H
This item is not returnable.
View return policy
Description
MAGEB18 Monoclonal specifically detects MAGEB18 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MAGEB18 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| MAGEB18 | |
| Full length recombinant protein of human MAGEB18 (NP_775970) produced in HEK293T cells. | |
| 0.1 mL | |
| 286514.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1H2 | |
| Western Blot, Immunohistochemistry | |
| MAGE-B18 antigen, melanoma antigen family B, 18, melanoma-associated antigen B18, MGC33889 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion