missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB18 Antibody (OTI1F5), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72575PEATT594
This item is not returnable.
View return policy
Description
MAGEB18 Monoclonal specifically detects MAGEB18 in Human samples. It is validated for Flow Cytometry.
Specifications
| MAGEB18 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| MAGEB18 | |
| Full length recombinant protein of human MAGEB18 (NP_775970) produced in HEK293T cells. | |
| 0.1 mL | |
| 286514.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI1F5 | |
| Flow Cytometry | |
| MAGE-B18 antigen, melanoma antigen family B, 18, melanoma-associated antigen B18, MGC33889 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction