missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£342.00 - £470.00
Specifications
| Antigen | MAGEB1 |
|---|---|
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18447951
|
Novus Biologicals
NBP1-85405-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18256597
|
Novus Biologicals
NBP1-85405 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAGEB1 Polyclonal specifically detects MAGEB1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MAGEB1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 4112 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LKVAHATAAEKEECPSSSPVLGDTPTSSPAAGIPQKPQGAPPTTTAAAAVSCTESDEGAKCQGEENASFSQATTSTESSVKDPVAWEAGMLMHFILRKYKMREPIMKADMLKVVDEKYKDHFTEILNGASR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Cancer/testis antigen 3.1, CT3.1cancer/testis antigen family 3, member 1, DAM10MAGE-B1 antigen, DSS/AHC critical interval MAGE superfamily 10, DSS-AHC critical interval MAGE superfamily 10, MAGEL1MAGE-XP antigen, MAGE-Xp, MAGEXP, melanoma antigen family B, 1, melanoma-associated antigen B1, MGC9322 | |
| MAGEB1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title