missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | MAGEA8 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
MAGEA8 Polyclonal specifically detects MAGEA8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MAGEA8 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Cancer/testis antigen 1.8, CT1.8MAGE-8 antigen, MAGE8cancer/testis antigen family 1, member 8, melanoma antigen family A, 8, melanoma-associated antigen 8, MGC2182 | |
| MAGEA8 | |
| IgG | |
| 35 kDa |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Q9BUN9 | |
| 4107 | |
| Synthetic peptides corresponding to MAGEA8(melanoma antigen family A, 8) The peptide sequence was selected from the N terminal of MAGEA8. Peptide sequence EEVTDSGSPSPPQSPEGASSSLTVTDSTLWSQSDEGSSSNEEEGPSTSPD. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title