missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84152-0.1ML
This item is not returnable.
View return policy
Description
MAGEA6 Polyclonal specifically detects MAGEA6 in Human samples. It is validated for Western Blot.
Specifications
| MAGEA6 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| CT1.6MGC52297, MAGE-3b, MAGE3B, MAGE3B antigen, MAGE6member 6, melanoma antigen family A, 6, melanoma-associated antigen 6 | |
| The immunogen is a synthetic peptide directed towards the C terminal region of human MAGEA6 | |
| 0.1 mL | |
| Cancer | |
| 4105 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction