missing translation for 'onlineSavingsMsg'
Learn More

MAGEA4 Antibody (OTI2C1), DyLight 594, Novus Biologicals™

Product Code. 18401345 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401345 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401345 Supplier Novus Biologicals Supplier No. NBP272571DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAGEA4 Monoclonal antibody specifically detects MAGEA4 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEA4
Applications Western Blot, Immunofluorescence, Immunofluorescence
Classification Monoclonal
Clone OTI2C1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias MAGE-X2 antigen, melanoma antigen family A, 4, melanoma-associated antigen 4, member 4
Host Species Mouse
Immunogen Full length recombinant protein of human MAGEA4 (NP_001011550) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4103
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.