missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1A9F3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60054-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita.Specifications
| MAGEA3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P43357 | |
| Magea3 | |
| MAGEA3 Fusion Protein Ag0265 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1A9F3 | |
| Unconjugated | |
| MAGEA3 | |
| Antigen MZ2 D, Cancer/testis antigen 1.3, CT1.3, HIP8, HYPD, MAGE 3 antigen, MAGE3, MAGEA3, MAGE-A3, MAGEA6, melanoma antigen family A, 3, Melanoma associated antigen 3 | |
| Mouse | |
| Protein G | |
| RUO | |
| 17139, 4102 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction