missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGEA3 Monoclonal Antibody (OTI2B1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526491
This item is not returnable.
View return policy
Description
MAGEA3 Monoclonal Antibody for Western Blot
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita.
Specifications
| MAGEA3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P43357 | |
| MAGEA3 | |
| Full length human recombinant protein of MAGEA3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI2B1 | |
| Unconjugated | |
| MAGEA3 | |
| Antigen MZ2-D; cancer/testis antigen 1.3; cancer/testis antigen family 1, member 3; CT1.3; HIP8; HYPD; MAGE family member A3; MAGE3; MAGE-3 antigen; MAGEA3; Mage-a3; MAGEA6; melanoma antigen family A, 3; melanoma antigen family A3; melanoma antigen, family A, 3; melanoma-associated antigen 3; MGC14613 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4102 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction