Learn More
MAGEA3 Antibody (CT Atlantic patent anti-MAGE-A3) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28060JF585
Product Code. 30163680
  • £328.00 / 0.10mL
Description
MAGEA3 Monoclonal antibody specifically detects MAGEA3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MAGEA3
Monoclonal
Janelia Fluor 585
Antigen MZ2-D, Cancer/testis antigen 1.3, CT1.3 MAGE-3 antigen, HIP8, MAGE3 MAGEA6, melanoma antigen family A, 3, melanoma-associated antigen 3, member 3, MGC14613
MAGEA3
0.1 mL
Cancer
4102
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
CT Atlantic patent anti-MAGE-A3
50mM Sodium Borate
Human
Protein A purified
RUO
Primary
Human
Purified