missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA3 Antibody (CT Atlantic patent anti-MAGE-A3), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28060CL1
This item is not returnable.
View return policy
Description
MAGEA3 Monoclonal antibody specifically detects MAGEA3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| MAGEA3 | |
| Monoclonal | |
| CoraFluor 1 | |
| Antigen MZ2-D, Cancer/testis antigen 1.3, CT1.3 MAGE-3 antigen, HIP8, MAGE3 MAGEA6, melanoma antigen family A, 3, melanoma-associated antigen 3, member 3, MGC14613 | |
| MAGEA3 | |
| 0.1 mL | |
| Cancer | |
| 4102 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| CT Atlantic patent anti-MAGE-A3 | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction