missing translation for 'onlineSavingsMsg'
Learn More

MAGEA12 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18389090 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389090 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389090 Supplier Novus Biologicals Supplier No. NBP2841510.1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAGEA12 Polyclonal specifically detects MAGEA12 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen MAGEA12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias Cancer/testis antigen 1.12, CT1.12MAGE12F antigen, MAGE-12 antigen, MAGE12cancer/testis antigen family 1, member 12, melanoma antigen family A, 12, melanoma-associated antigen 12
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of MAGEA12
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4111
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.