missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA11 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04410-100UL
This item is not returnable.
View return policy
Description
MAGEA11 Polyclonal specifically detects MAGEA11 in Human samples. It is validated for Western Blot.
Specifications
| MAGEA11 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4110 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Cancer/testis antigen 1.11, CT1.11MAGE-11 antigen, MAGE11member 11, MAGEA-11, melanoma antigen family A, 11, melanoma-associated antigen 11, MGC10511 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human MAGEA11 (NP_005357.2). | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction