missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- The MagBeads FastRNA Kit for Feces is a scalable and automatable magnetic bead-based RNA purification technology, designed for the efficient isolation of total RNA from fecal samples.
- Available in prefilled formats – Ready-to-use for MPure-32™ (Cat. No. 117040300) and Ready-to-use for MPure-96™ (Cat. No. 117040400) aNAP Systems.
- The composition of fecal samples is largely dependent on diet, which includes fibers, undigested particles, bilirubin, complex polysaccharides, and lipids. These compounds impair sample homogenization, decreasing both the quantity and quality of the extracted RNA. The MagBeads FastRNA Kit for Feces integrates our
- state-of-the-art beads-beating technology, a lysis chemistry compatible with the widest range of samples and our proprietary inhibitor removal expertise. A new technology of selective binding of RNA along with our FastDNase I Enzyme treatment allows the complete removal of DNA. The MagBeads FastRNA Kit for Feces streamlines RNA purification by overcoming the challenges associated with diverse fecal sample types, making the process both hassle-free and time efficient. A DNase I treatment step effectively removes contaminated DNA, ensuring high RNA yield and purity. The extracted RNA is immediately compatible with downstream applications such as RT-qPCR amplification, eliminating the need for additional inhibitor or DNA removal steps.
Specifications
Specifications
| Buffer | Lysis Buffer |
| Control Type | RNA Control |
| Isolation Technology | Magnetic Bead |
| Species | All |
| Sample Type | Feces |
| Product Type | MagBeads FastRNA Kit |
| Sterility | Non-sterile |
| Target | Total RNA |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?