missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP BIOMEDICALS MagBeads FastDNA Kit for FFPE Ready-to-Use for MPure-32 96 Preps
Shop All MP Biomedicals ProductsDescription
MagBeads FastDNA™ Kit for FFPE is based on the purification method of high binding magnetic particles.
The sample is lysed and digested. DNA is released into the lysate. After addition of magnetic particles and binding solution:
- DNA will be adsorbed on the surface of magnetic particles
- Impurities such as proteins will be removed without adsorption
The adsorbed particles were washed with:
- Washing buffer to remove the proteins and impurities
- Ethanol to remove salts
- And finally the DNA was eluted with Elution Buffer
Specifications
Specifications
| Format | Kit |
| Isolation Technology | Magnetic Bead |
| For Use With (Application) | DNA Extraction |
| Product Type | DNA Kit for FFPE |
Safety and Handling
Product Identifier
- (MPure-32) MagBeads FastDNA Kit for FFPE
- Danger
- Acute toxicity Category 4
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin corrosion/irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H304-May be fatal if swallowed and enters airways.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sodium Dodecyl Sulphate, Proteinase K, Guanidine Hydrochloride, Ethanol, Guanidine Thiocyanate , Tri
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction