missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ MagBeads FastDNA™ Kit for Feces
Shop All MP Biomedicals Products
Click to view available options
:
MagBeads FastDNA™ Kit for Feces
Description
- Quick and efficient isolation of high-quality genomic DNA from:
- Fresh or frozen human and animal feces, intestinal contents
- < 60 min. for extraction
- High yield and throughput
- Free from inhibitors
- Fresh or frozen human and animal feces, intestinal contents
- Samples placed into Lysing Matrix E tubes and used with FastPrep™ instruments to lyse within 40 seconds:
- Host cells
- Bacteria
- Fungi
- Viruses
- Protists
- Other cells in fecal samples
Kit:
- Compatible with most automated nucleic acid extraction instruments
- Can be operated manually
- No phenol/chloroform or other toxic chemicals
- Consists of specially formulated reagents to eliminate:
- Humic acid
- Polysaccharides
- Phenolic compounds
- Enzyme inhibitors from feces
- Allows for extraction of highly pure genomic DNA ready for:
- PCR
- Restriction digestion
- Electrophoresis
Specifications
Specifications
| Detection Method | Magnetic Extraction |
| Isolation Technology | Magnetic Particles |
| Sample Type | Feces |
| For Use With (Equipment) | Homogenizer such as FastPrep™-24 5G for automated extractions |
| For Use With (Application) | DNA Extraction |
| No. of Reactions | 50 Preps |
| Product Type | DNA Extraction Kit |
| Target | DNA |
| Test Time | <60 min. |
Safety and Handling
Product Identifier
- MagBeads FastDNA Kit For Feces
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Guanidine Thiocyanate, Acetic Acid, Lithium Chloride, SodiumDodecyl Sulphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction