Learn More
MP BIOMEDICALS Magbeads FastDNA Kit for Feces Ready-to-Use for MPure-32 96 Preps
Shop All MP Biomedicals ProductsDescription
The MagBeads FastDNA™ Kit for Feces allows quick and efficient isolation of high-quality genomic DNA from fresh or frozen human and animal feces in less than 60 mins. Samples are placed into Lysing Matrix E tube and used with FastPrep™ Instruments from MP Biomedicals to effectively lyse within 40 seconds:
- host cells
- bacteria
- fungi
- viruses
- protists
- and other cells present in fecal samples.
The kit is also compatible with most of the automated nucleic acid extraction instruments on the market or it can be operated manually. It consists of specially formulated reagents to eliminate:
- humic acid
- polysaccharides
- phenolic compounds
- enzyme inhibitors from feces
And thus allow extraction of highly pure genomic DNA ready for:
- PCR
- restriction digestion
- electrophoresis
- other desired applications.
Usage Statement:
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Specifications
| Format | Kit |
| Isolation Technology | Magnetic Bead |
| For Use With (Application) | DNA Purification |
| Product Type | DNA Kit for Feces |
Safety and Handling
- (MPure-32) MagBeads FastDNA Kit for Feces
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sodium Dodecyl Sulphate, Ethanol, Isopropanol, Guanidine Thiocyanate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.