missing translation for 'onlineSavingsMsg'
Learn More

MP BIOMEDICALS MagBeads FastDNA Kit for Blood Ready-to-Use for MPure-32 96 Preps

Product Code. 30067482 Shop All MP Biomedicals Products
This item is not returnable. View return policy

Product Code. 30067482

Brand: MP BIOMEDICALS 117033700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Magbeads FastDNA™ Kit for Blood is intended for purification of total DNA from whole blood, plasma, serum, buffy coat, bone marrow, other body fluids, lymphocytes, or cultured cells using the MPure-32™ aNAP System.

Magbeads FastDNA™ Kit for Blood is based on the purification method of high binding magnetic particles.

  • The sample is lysed and digested.
  • DNA is released into the lysate.
  • After addition of magnetic particles and binding solution, DNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption.
  • The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts, and finally the DNA was eluted with Elution Buffer.
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
For Use With (Application) DNA Purification
Product Type DNA Kit for Blood
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (MPure-32) MagBeads FastDNA Kit for Blood
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Proteinase K, Guanidine Hydrochloride, Guanidine Thiocyanate, Isopropanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.