missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Magbeads Fast Circulating DNA Kit

Product Code. 17782448 Shop All MP Biomedicals Products
Click to view available options
:
192 Preps
This item is not returnable. View return policy

Product Code. 17782448

Brand: MP Biomedicals™ 116577192

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MagBeads Fast Circulating DNA Kit is intended for purification of high-quality circulating DNA (cfDNA) from cell-free body fluids (such as plasma, serum). The purified DNA is suitable for direct use in downstream applications such as PCR, real-time PCR, Biochip analysis and NGS.

MagBeads Fast Circulating DNA Kit is based on the purification method of high binding magnetic particles:

  • The sample is lysed and digested
  • DNA is released into the lysate
  • After addition of magnetic particles and binding solution, DNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption
  • The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts
  • Finally the DNA was eluted with Elution Buffer
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
Scale Mini
Sample Type Cell-Free Body Fluids
Sequencing Type DNA and RNA Sequencing
No. of Reactions 192 Preps
compliance-icons compliance-icons
Product Identifier
  • MagBeads Fast Circulating DNA Kit
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Proteinase K
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.