missing translation for 'onlineSavingsMsg'
Learn More

Magainin 1, MedChemExpress

Product Code. 16491918
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491918 5 mg 5mg
16479428 10 mg 10mg
16412739 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491918 Supplier Medchem Express Supplier No. HYP0269

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Magainin 1 (Magainin I) is an antimicrobial and amphipathic peptide isolated from the skin of Xenopus laevis. Magainin 1 exhibits antibiotic activity against numerous Gram-negative and Gram-positive bacteria.

Magainin 1 (Magainin I) is an antimicrobial and amphipathic peptide isolated from the skin of Xenopus laevis. Magainin 1 exhibits antibiotic activity against numerous Gram-negative and Gram-positive bacteria.

Specifications

Chemical Name or Material Magainin 1
Physical Form Powder
CAS 108433-99-4
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C112H177N29O28S
For Use With (Application) COVID-19-immunoregulation
Synonym Magainin I
SMILES [GIGKFLHSAGKFGKAFVGEIMKS]
Molecular Weight (g/mol) 2409.85
Formula Weight 2409.85
Percent Purity 99.66%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.