missing translation for 'onlineSavingsMsg'
Learn More

MafK Antibody (2F7), DyLight 405, Novus Biologicals™

Product Code. 18404396 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404396 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404396 Supplier Novus Biologicals Supplier No. NBP222570V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MafK Monoclonal antibody specifically detects MafK in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MafK
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2F7
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias FLJ32205, MGC71717, transcription factor MafK, ubiquitous (p18), v-maf avian musculoaponeurotic fibrosarcoma oncogene family, protein K, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene family, protein K, v-maf musculoaponeurotic fibrosarcoma oncogene homolog K (avian)
Host Species Mouse
Immunogen Recombinant human MAFK (1-156aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 7975
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.