missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MafG Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82278-0.1ML
This item is not returnable.
View return policy
Description
MafG Polyclonal specifically detects MafG in Mouse samples. It is validated for Western Blot.
Specifications
| MafG | |
| Polyclonal | |
| PBS, 2% sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 4097 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| basic leucine zipper transcription factor MafG, hMAF, MGC13090, MGC20149, transcription factor MafG, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene family, protein G, V-maf musculoaponeurotic fibrosarcoma oncogene homolog G, v-maf musculoaponeurotic fibrosarcoma oncogene homolog G (avian) | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Mouse Mafg | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction