missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAFG Monoclonal Antibody (370412)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561362

Product Code. 30535300

  • £382.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Globin gene expression is regulated through nuclear factor erythroid-2 (NFE2) elements located in enhancer-like locus control regions positioned many kb upstream of alpha- and beta-gene clusters (summarized by Blank et al., 1997 [PubMed 9166829]). NFE2 DNA-binding activity consists of a heterodimer containing a ubiquitous small Maf protein (MafF, MIM 604877; MafG; or MafK, MIM 600197) and the tissue-restricted protein p45 NFE2 (MIM 601490). Both subunits are members of the activator protein-1-like superfamily of basic leucine zipper (bZIP) proteins (see MIM 165160).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAFG
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
O15525
MAFG
E. coli-derived recombinant human MafG, Met1-Ser162.
100 μg
Primary
Human
Antibody
IgG2a
Western Blot
370412
Unconjugated
MAFG
AA545192; avian musculoaponeurotic fibrosarcoma virus (v-maf) AS42 oncogene, protein G; basic leucine zipper transcription factor MafG; C630022N07Rik; hMAF; MAF bZIP transcription factor G; MAFG; MGC13090; MGC20149; Transcription factor MafG; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog G; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein G; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein G (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog G
Mouse
Protein A/G
RUO
4097
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.