Learn More
Invitrogen™ MAFF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585462
Description
Keep as concentrated solution. Predicted reactivity: Mouse (97%), Rat (97%), Chicken (86%), Bovine (99%). Positive Control: A431, A431 nuclear extract, rat skin, Huh7, HepG2, human MAFF-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a basic leucine zipper (bZIP) transcription factor that lacks a transactivation domain. It is known to bind the US-2 DNA element in the promoter of the oxytocin receptor (OTR) gene and most likely heterodimerizes with other leucine zipper-containing proteins to enhance expression of the OTR gene during term pregnancy. The encoded protein can also form homodimers, and since it lacks a transactivation domain, the homodimer may act as a repressor of transcription. This gene may also be involved in the cellular stress response. Multiple transcript variants encoding two different isoforms have been found for this gene.
Specifications
| MAFF | |
| Polyclonal | |
| Unconjugated | |
| Maff | |
| hMafF; hypothetical protein MGC63868; LOW QUALITY PROTEIN: transcription factor MafF; MAF bZIP transcription factor F; Maff; Transcription factor MafF; transcription factor mafT; U-Maf; v-maf avian musculoaponeurotic fibrosarcoma oncogene family protein F; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog F; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein F; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein F (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog f; zgc:63868 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23764, 366960 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9ULX9 | |
| Maff | |
| Recombinant protein encompassing a sequence within the center region of human MAFF. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.