missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAFF Polyclonal Antibody

Description
Immunogen sequence: LSSKALKIKRE LSENTPHLSD EALMGLSVRE L Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
The protein encoded by this gene is a basic leucine zipper (bZIP) transcription factor that lacks a transactivation domain. It is known to bind the US-2 DNA element in the promoter of the oxytocin receptor (OTR) gene and most likely heterodimerizes with other leucine zipper-containing proteins to enhance expression of the OTR gene during term pregnancy. The encoded protein can also form homodimers, and since it lacks a transactivation domain, the homodimer may act as a repressor of transcription. This gene may also be involved in the cellular stress response. Multiple transcript variants encoding two different isoforms have been found for this gene.
Specifications
Specifications
| Antigen | MAFF |
| Applications | ChIP Assay, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Maff |
| Gene Accession No. | Q9ULX9 |
| Gene Alias | hMafF; hypothetical protein MGC63868; LOW QUALITY PROTEIN: transcription factor MafF; MAF bZIP transcription factor F; Maff; Transcription factor MafF; transcription factor mafT; U-Maf; v-maf avian musculoaponeurotic fibrosarcoma oncogene family protein F; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog F; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein F; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein F (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog f; zgc:63868 |
| Gene Symbols | Maff |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?