missing translation for 'onlineSavingsMsg'
Learn More

Mafenide Hydrochloride, Mikromol™

Product Code. 17953796
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17953796 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17953796 Supplier Mikromol Supplier No. MM3127.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Mafenide Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mafenide Hydrochloride
CAS 138-37-4
Analyte or Component Names Mafenide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C7 H10 N2 O2 S . Cl H
Certifications/Compliance ISO 17025
Synonym Benzenesulfonamide, 4-(aminomethyl)-, hydrochloride (1:1), Benzenesulfonamide, 4-(aminomethyl)-, monohydrochloride (9CI), 4-(Aminomethyl)benzene sulfonamide monohydrochloride, 4-Aminosulfonylbenzylamine hydrochloride, 4-Homosulfanilamide hydrochloride, Homosulfamine, Homosulfamine hydrochloride, Homosulfanilamide hydrochloride, Mafenide hydrochloride, Marfanil, Sulfamylon hydrochloride, p-(Aminomethyl)benzenesulfonamide hydrochloride, p-(Aminomethyl)benzenesulfonamide monohydrochloride, p-Aminomethylphenylsulfonamide hydrochloride, p-Toluenesulfonamide, alpha-amino-, hydrochloride (6CI), p-Toluenesulfonamide, alpha-amino-, monohydrochloride (8CI), alpha-Amino-p-toluenesulfonamide hydrochloride
InChI Formula InChI=1S/C7H10N2O2S.ClH/c8-5-6-1-3-7(4-2-6)12(9,10)11;/h1-4H,5,8H2,(H2,9,10,11);1H
SMILES Cl.NCc1ccc(cc1)S(=O)(=O)N
IUPAC Name 4-(aminomethyl)benzenesulfonamide;hydrochloride
Molecular Weight (g/mol) 222.69
Quantity 100 mg
Formula Weight 222.023 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.