missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAF1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA523712

Product Code. 13298788

  • £439.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is predicted to react with bovine and rat based on sequence homology.

This gene encodes a protein that is similar to Maf1, a Saccharomyces cerevisiae protein highly conserved in eukaryotic cells. Yeast Maf1 is a negative effector of RNA polymerase III (Pol III). It responds to changes in the cellular environment and represses pol III transcription. Biochemical studies identified the initiation factor TFIIIB as a target for Maf1-dependent repression.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAF1
Polyclonal
Unconjugated
MAF1
1110068E11Rik; AU042856; homolog of yeast MAF1; MAF1; MAF1 homolog (S. cerevisiae); MAF1 homolog, negative regulator of RNA polymerase III; MAF1 negative regulator of RNA polymerase III; repressor of RNA polymerase III transcription MAF1 homolog
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
68877, 84232
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9D0U6, Q9H063
MAF1
KLH conjugated synthetic peptide between 90-117 amino acids from the central region of human MAF1.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.