missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555195
This item is not returnable.
View return policy
Description
Immunogen sequence: MKLLENSSFE AINSQLTVET GDAHIIGRIE SYSCKMAGDD KHMFKQFCQE GQPHVLEALS PPQTSGLS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a protein that is similar to Maf1, a Saccharomyces cerevisiae protein highly conserved in eukaryotic cells. Yeast Maf1 is a negative effector of RNA polymerase III (Pol III). It responds to changes in the cellular environment and represses pol III transcription. Biochemical studies identified the initiation factor TFIIIB as a target for Maf1-dependent repression.
Specifications
| MAF1 | |
| Polyclonal | |
| Unconjugated | |
| MAF1 | |
| 1110068E11Rik; AU042856; homolog of yeast MAF1; MAF1; MAF1 homolog (S. cerevisiae); MAF1 homolog, negative regulator of RNA polymerase III; MAF1 negative regulator of RNA polymerase III; repressor of RNA polymerase III transcription MAF1 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84232 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H063 | |
| MAF1 | |
| Recombinant protein corresponding to Human MAF1. Recombinant protein control fragment (Product #RP-93931). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction