missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55013-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to MAF.
The protein encoded by this gene is an integral plasma membrane protein which promotes cell adhesion. The encoded protein is thought to undergo O-linked glycosylation. Expression of this gene has been shown to be induced by gamma interferon in some cancer cells. Two transcript variants encoding the same protein have been found for this gene.Specifications
| MAF | |
| Polyclonal | |
| Unconjugated | |
| MAF | |
| c MAF, cMAF, MAF, Proto oncogene c Maf, Transcription factor Maf | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17132, 4094, 54267 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75444, P54843, P54844 | |
| Maf | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction