missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAEL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26666-1-AP-150UL
This item is not returnable.
View return policy
Description
The mammalian homolog of the Drosophila protein Maelstrom is expressed in the male germline and localizes to the sex body in spermatocytes and the chromatid body in round spermatids. Similar to its expression in Drosophila, Maelstrom is a component of nuages, a germ-cell specific organelle and is thought to be essential for spermatogenesis and transposon repression during meiosis. In humans, Maelstrom has been found to be expressed only in the testis and in various cancer cell lines. Treatment of these cell lines with the demethylating agent 5′-Aza-2-Deoxycytidine significantly upregulated Maelstrom levels, indicating that its expression is regulated by DNA methylation.Specifications
| MAEL | |
| Polyclonal | |
| Unconjugated | |
| MAEL | |
| MAEL, maelstrom homolog (Drosophila), Protein maelstrom homolog, RP11 102C16.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 364039, 84944, 98558 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BVN9, Q96JY0 | |
| Mael | |
| MAEL Fusion Protein Ag24524 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction