missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MADD Recombinant Rabbit Monoclonal Antibody (24GB110)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557457
This item is not returnable.
View return policy
Description
MADD Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Tumor necrosis factor receptor 1 (TNFR1) contains a death receptor domain that mediates interactions involved in downstream signaling of TNF. A novel protein, MADD (mitogen activated protein (MAP)-kinase activating death domain protein) has recently been shown to associate with the death domain of TNFR1 through its own C-terminal death domain. This interaction is critical for TNFR1 signal generation. Overexpression of MADD activates the MAP (Mitogen Activated Protein) kinases extracellular signal-related kinase (ERK) and c-Jun N-terminal kinase (JNK) and induces the phosphorylation of cytosolic phospholipase A2. This discovery suggests that MADD links tumor necrosis factor TNF and MAP kinase activation and arachidonic acid release. MADD has a 98% homology with another protein, differentially expressed in neoplasmic vs. normal cells (DENN). DENN is a substrate for JNK3, and is important in the hypoxia and stress induced intracellular signalling pathways. MADD also has a 93% sequence conservation with a GDP/GTP exchange protein, Rab3.
Specifications
| MADD | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O08873, Q80U28, Q8WXG6 | |
| MADD | |
| A synthesized peptide derived from human DENN (aa 1611-1647). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB110 | |
| Unconjugated | |
| MADD | |
| 9630059K23Rik; DENN; differentially expressed in normal and neoplastic cells; IG20; Insulinoma glucagonoma clone 20; insulinoma-glucagonoma protein 20; Kiaa0358; Madd; MAP kinase activating death domain; MAP kinase-activating death domain protein; MAP-kinase activating death domain; rab3 GDP/GTP exchange factor; Rab3 GDP/GTP exchange protein; Rab3 GEP; RAB3GEP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 228355, 8567, 94193 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction