missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MADD Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06250PE
This item is not returnable.
View return policy
Description
MADD Polyclonal antibody specifically detects MADD in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| MADD | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| DENNFLJ36300, Differentially expressed in normal and neoplastic cells, FLJ35600, IG20, Insulinoma glucagonoma clone 20, insulinoma-glucagonoma protein 20, KIAA0358RAB3GEP, MAP kinase-activating death domain protein, MAP-kinase activating death domain, Rab3 GDP/GTP exchange factor | |
| Produced in rabbits immunized with E. coli-derived Human MADD fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 8567 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction