missing translation for 'onlineSavingsMsg'
Learn More

MADD Antibody, Novus Biologicals™

Product Code. 18412402 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412402 25ul 25µL
18298006 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18412402 Supplier Novus Biologicals Supplier No. NBP18525825ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MADD Polyclonal specifically detects MADD in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MADD
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias DENNFLJ36300, Differentially expressed in normal and neoplastic cells, FLJ35600, IG20, Insulinoma glucagonoma clone 20, insulinoma-glucagonoma protein 20, KIAA0358RAB3GEP, MAP kinase-activating death domain protein, MAP-kinase activating death domain, Rab3 GDP/GTP exchange factor
Gene Symbols MADD
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:YSQQINEVLDQLANLNGRDLSIWSSGSRHMKKQTFVVHAGTDTNGDIFFMEVCDDCVVLRSNIGTVYERWWYEKLINMTYCPKTKVLCLWRRNGSETQ
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8567
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.