missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MADD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77033SS
This item is not returnable.
View return policy
Description
MADD Polyclonal specifically detects MADD in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| MADD | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| DENNFLJ36300, Differentially expressed in normal and neoplastic cells, FLJ35600, IG20, Insulinoma glucagonoma clone 20, insulinoma-glucagonoma protein 20, KIAA0358RAB3GEP, MAP kinase-activating death domain protein, MAP-kinase activating death domain, Rab3 GDP/GTP exchange factor | |
| Rabbit | |
| 200-220 kDa | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/ml | |
| Western Blot 1-2mg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 ug/ml | |
| AAD12154 | |
| MADD | |
| Antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human MADD. The immunogen is located within the last 50 amino acids of MADD. Amino Acid Squence: AAVHSSEEDLRTPPRPVSS | |
| Ion exchange chromatography | |
| Signal Transduction | |
| 8567 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction