missing translation for 'onlineSavingsMsg'
Learn More

MADD Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18393146 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393146 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393146 Supplier Novus Biologicals Supplier No. NBP306250AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MADD Polyclonal specifically detects MADD in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MADD
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DENNFLJ36300, Differentially expressed in normal and neoplastic cells, FLJ35600, IG20, Insulinoma glucagonoma clone 20, insulinoma-glucagonoma protein 20, KIAA0358RAB3GEP, MAP kinase-activating death domain protein, MAP-kinase activating death domain, Rab3 GDP/GTP exchange factor
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MADD fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8567
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.