missing translation for 'onlineSavingsMsg'
Learn More

MAdCAM1 Monoclonal Antibody (MECA-367), Functional Grade, eBioscience™, Invitrogen™

Product Code. 17111432
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17111432 50 mg 50mg
17121432 5 mg 5mg
17101432 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17111432 Supplier Affymetrix eBioscience Supplier No. 165997050

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rat Monoclonal Antibody

The MECA-367 monoclonal antibody reacts with mouse mucosal vascular addressin-1 (MAdCAM-1), a 50-60 kDa member of the Ig superfamily. This antigen is expressed in mucosal lymphoid tissue and lamina propria and is a counter-receptor for CD62L and CD49d. MAdCAM-1 is involved in homing of subpopulation of memory T cells to the gut lamina propria, and recirculation of naive lymphocytes in Peyer's patches and mesenteric lymph nodes. Applications Reported: MECA-367 can be used in flow cytometric analysis, immunohistochemical staining of frozen mouse tissue sections, blocking in adhesion assays, immunoprecipitation and immunoblotting (WB). Applications Tested: The MECA-367 antibody has been tested by flow cytometric analysis of mouse bone marrow cells and splenocytes and can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in other applications. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

MAdCAM1 Monoclonal antibody specifically detects MAdCAM1 in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAdCAM1
Applications Flow Cytometry, Functional Assay, Neutralization
Classification Monoclonal
Clone MECA-367
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene MADCAM1
Gene Accession No. Q61826
Gene Alias AV211525; hMAdCAM-1; MACAM1; Madcam1; MAdCAM-1; mMAdCAM-1; mucosal addressin cell adhesion molecule 1; mucosal addressin cell adhesion molecule-1; mucosal vascular addressin cell adhesion molecule 1; rMAdCAM-1
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17123
Target Species Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.