missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAdCAM1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21917-1-AP-150UL
This item is not returnable.
View return policy
Description
MAdCAM-1 protein is an endothelial cell adhesion molecule that interacts preferentially with the leukocyte beta7 integrin LPAM-1 (alpha4beta7), L-selectin, and VLA-4 (alpha4beta1) on myeloid cells to direct leukocytes into mucosal and inflamed tissues. It is a member of the immunoglobulin family and is similar to ICAM1 and VCAM1. At least seven alternatively spliced transcripts encoding different protein isoforms have been found for this gene, but the full-length nature of some variants has not been determined.Specifications
| MAdCAM1 | |
| Polyclonal | |
| Unconjugated | |
| MADCAM1 | |
| hMAdCAM 1, MACAM1, MAdCAM1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8174 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13477 | |
| MADCAM1 | |
| MAdCAM1 Fusion Protein Ag16184 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction