missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAdCAM1 Mouse anti-Human, Biotin, Clone: 2A12E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech BIOTIN-66594-100UL
This item is not returnable.
View return policy
Description
MAdCAM1 Monoclonal antibody apecifically detetects MAdCAM1 antigen in Human Samples. It is validated for Immunohistochemistry.Specifications
| MAdCAM1 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| MADCAM1 | |
| MAdCAM1 fusion protein Ag16184 | |
| 100 μL | |
| 8174 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG2a |
| Immunohistochemistry | |
| 2A12E8 | |
| Biotin | |
| BC142629 | |
| Mouse | |
| Protein A purification | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction