missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAdCAM-1 Recombinant Rabbit Monoclonal Antibody (24GB660)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557055
This item is not returnable.
View return policy
Description
MAdCAM-1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is an endothelial cell adhesion molecule that interacts preferentially with the leukocyte beta7 integrin LPAM-1 (alpha4beta7), L-selectin, and VLA-4 (alpha4beta1) on myeloid cells to direct leukocytes into mucosal and inflamed tissues. It is a member of the immunoglobulin family and is similar to ICAM1 and VCAM1. At least seven alternatively spliced transcripts encoding different protein isoforms have been found for this gene, but the full-length nature of some variants has not been determined.
Specifications
| MAdCAM-1 | |
| Recombinant Monoclonal | |
| 1.51 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13477 | |
| MADCAM1 | |
| A synthesized peptide derived from human MADCAM1 (aa 1-53). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB660 | |
| Unconjugated | |
| MADCAM1 | |
| AV211525; hMAdCAM-1; MACAM1; MADCAM1; MAdCAM-1; mMAdCAM-1; mucosal addressin cell adhesion molecule 1; mucosal addressin cell adhesion molecule-1; mucosal vascular addressin cell adhesion molecule 1; rMAdCAM-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8174 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction