missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAdCAM-1 Rat anti-Mouse, BUV615, Clone: MECA-89, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 751176
This item is not returnable.
View return policy
Description
MAdCAM-1 Monoclonal antibody specifically detects MAdCAM-1 in Mouse samples. It is validated for Flow Cytometry
Specifications
| MAdCAM-1 | |
| Monoclonal | |
| Brilliant Ultraviolet 615 | |
| MAdCAM-1; Madcam1; mMAdCAM-1; MADCA | |
| Mouse mesenteric and peripheral lymph node cells | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| MECA-89 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction