missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MAdCAM-1 Monoclonal antibody specifically detects MAdCAM-1 in Mouse samples. It is validated for Western Blot, Neutralization
Specifications
Specifications
| Dilution | Western Blot, Block/Neutralize |
| Formulation | PBS |
| Gene Alias | hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1 |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Gene ID (Entrez) | 8174 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?