missing translation for 'onlineSavingsMsg'
Learn More

MAdCAM-1 Antibody (RM0284-11A55), DyLight 594, Novus Biologicals™

Product Code. 18420818 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420818 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420818 Supplier Novus Biologicals Supplier No. NBP211785DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

MAdCAM-1 Monoclonal antibody specifically detects MAdCAM-1 in Mouse samples. It is validated for Western Blot, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAdCAM-1
Applications Western Blot, Neutralization
Classification Monoclonal
Clone RM0284-11A55
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1
Host Species Rat
Immunogen Mouse recombinant MAdCAM-1 (extracellular domain)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8174
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.