Learn More
MAdCAM-1 Antibody (RM0284-11A55), Alexa Fluor™ 647, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-11785AF647
Product Code. 18141080
  • £476.00 / 0.10mL
Description
MAdCAM-1 Monoclonal specifically detects MAdCAM-1 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MAdCAM-1
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
MADCAM1
Mouse recombinant MAdCAM-1 (extracellular domain)
0.1 mL
8174
Store at 4C in the dark.
IgG2
Western Blot
RM0284-11A55
Western Blot
hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1
Rat
Protein A or G purified
Primary
Mouse
Purified