missing translation for 'onlineSavingsMsg'
Learn More

MAdCAM-1 Antibody (314G8), Novus Biologicals™

Product Code. 18349882 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349882 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349882 Supplier Novus Biologicals Supplier No. NBP311286

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAdCAM-1 Monoclonal specifically detects MAdCAM-1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAdCAM-1
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Immunoassay
Classification Monoclonal
Clone 314G8
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1
Host Species Mouse
Immunogen The monocolonal antibody 314G8 reacts with human mucosal addressin cell adhesion molecules-1 (MAdCAM-1)
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8174
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.