missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAdCAM-1 Antibody (12B5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26157-50ul
This item is not returnable.
View return policy
Description
MAdCAM-1 Monoclonal antibody specifically detects MAdCAM-1 in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| MAdCAM-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8174 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, ELISA | |
| 12B5 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1 | |
| A synthesized peptide derived from Human MAdCAM-1 [UniProt Q13477] | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction