missing translation for 'onlineSavingsMsg'
Learn More

MAD4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18344356 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344356 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344356 Supplier Novus Biologicals Supplier No. NBP310957100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAD4 Polyclonal specifically detects MAD4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS buffer, 2% sucrose
Gene Alias BHLHC12, bHLHc12Max-interacting transcriptional repressor MAD4, Class C basic helix-loop-helix protein 12, Mad4 homolog, MAD4Max-associated protein 4, MAX dimerization protein 4, Max dimerizer 4, MST149, MSTP149, MXD4 MAX dimerization protein 4
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N terminal region of human MAD4 (NP_006445). Peptide sequence SLLILLEAAEYLERRDREAEHGYASVLPFDGDFAREKTKAAGLVRKAPNN
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10608
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.