missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAD4 Monoclonal Antibody (OTI7F7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527071
This item is not returnable.
View return policy
Description
MAD4 Monoclonal Antibody for Western Blot, IHC (P)
This gene is a member of the MAD gene family. The MAD genes encode basic helix-loop-helix-leucine zipper proteins that heterodimerize with MAX protein, forming a transcriptional repression complex. The MAD proteins compete for MAX binding with MYC, which heterodimerizes with MAX forming a transcriptional activation complex. Studies in rodents suggest that the MAD genes are tumor suppressors and contribute to the regulation of cell growth in differentiating tissues.
Specifications
| MAD4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14582 | |
| MXD4 | |
| Full length human recombinant protein of MXD4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7F7 | |
| Unconjugated | |
| MXD4 | |
| 2810410A03Rik; bHLHc12; Class C basic helix-loop-helix protein 12; LOW QUALITY PROTEIN: max dimerization protein 4; MAD4; Mad4 homolog; max dimerization protein 4; Max dimerizer 4; max-associated protein 4; Max-interacting transcriptional repressor MAD4; MST149; MSTP149; MXD4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10608 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction