missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MAD4 Monoclonal antibody specifically detects MAD4 in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | MAD4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI7F7 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BHLHC12, bHLHc12Max-interacting transcriptional repressor MAD4, Class C basic helix-loop-helix protein 12, Mad4 homolog, MAD4Max-associated protein 4, MAX dimerization protein 4, Max dimerizer 4, MST149, MSTP149, MXD4 MAX dimerization protein 4 |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human MXD4(NP_006445) produced in E.coli. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?