missing translation for 'onlineSavingsMsg'
Learn More

MAD4 Antibody (OTI7F7), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18433495 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433495 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433495 Supplier Novus Biologicals Supplier No. NBP272567MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAD4 Monoclonal antibody specifically detects MAD4 in Human, Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD4
Applications Flow Cytometry
Classification Monoclonal
Clone OTI7F7
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias BHLHC12, bHLHc12Max-interacting transcriptional repressor MAD4, Class C basic helix-loop-helix protein 12, Mad4 homolog, MAD4Max-associated protein 4, MAX dimerization protein 4, Max dimerizer 4, MST149, MSTP149, MXD4 MAX dimerization protein 4
Host Species Mouse
Immunogen Full length human recombinant protein of human MXD4(NP_006445) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10608
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.