missing translation for 'onlineSavingsMsg'
Learn More

MAD4 Antibody, Novus Biologicals™

Product Code. 18396028 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396028 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396028 Supplier Novus Biologicals Supplier No. H00010608B02P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

MAD4 Polyclonal antibody specifically detects MAD4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_006445.1
Gene Alias BHLHC12, bHLHc12Max-interacting transcriptional repressor MAD4, Class C basic helix-loop-helix protein 12, Mad4 homolog, MAD4Max-associated protein 4, MAX dimerization protein 4, Max dimerizer 4, MST149, MSTP149, MXD4 MAX dimerization protein 4
Host Species Mouse
Immunogen MXD4 (NP_006445.1, 1 a.a. - 209 a.a.) full-length human protein. MELNSLLILLEAAEYLERRDREAEHGYASVLPFDGDFAREKTKAAGLVRKAPNNRSSHNELEKHRRAKLRLYLEQLKQLVPLGPDSTRHTTLSLLKRAKVHIKKLEEQDRRALSIKEQLQQEHRFLKRRLEQLSVQSVERVRTDSTGSAVSTDDSEQEVDIEGMEFGPGELDSVGSSSDADDHYSLQSGTGGDSGFGPHCRRLGRPALS
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10608
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.